Anti-SULT4A1 Mouse Monoclonal Antibody [clone: 634630]

Supplier: R & D Systems
MAB5826
102886-122EA 561.11 USD
102886-122
Anti-SULT4A1 Mouse Monoclonal Antibody [clone: 634630]
Antibodies
Type: Primary
Antigen: SULT4A1
Clonality: Monoclonal
Clone: 634630
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR