Anti-VAPA Mouse Monoclonal Antibody [clone: 604101]

Supplier: R & D Systems
MAB5820
220021-267EA 577.02 USD
220021-267
Anti-VAPA Mouse Monoclonal Antibody [clone: 604101]
Antibodies
Type: Primary
Antigen: VAPA
Clonality: Monoclonal
Clone: 604101
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR