Anti-IL22 Rat Monoclonal Antibody [clone: 140301]

Supplier: R & D Systems
MAB582
220021-266EA 763.57 USD
220021-266
Anti-IL22 Rat Monoclonal Antibody [clone: 140301]
Antibodies
Type: Primary
Antigen: IL22
Clonality: Monoclonal
Clone: 140301
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR