Anti-IL10 Mouse Monoclonal Antibody [clone: 155115]

Supplier: R & D Systems
MAB581
220021-264EA 763.57 USD
220021-264
Anti-IL10 Mouse Monoclonal Antibody [clone: 155115]
Antibodies
Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: 155115
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Cotton Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR