Anti-SOCS7 Mouse Monoclonal Antibody [clone: 519106]

Supplier: R & D Systems
MAB5809
220021-263EA 561.11 USD
220021-263
Anti-SOCS7 Mouse Monoclonal Antibody [clone: 519106]
Antibodies
Type: Primary
Antigen: SOCS7
Clonality: Monoclonal
Clone: 519106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR