Anti-BTK Mouse Monoclonal Antibody [clone: 566928]

Supplier: R & D Systems
MAB5807
220021-261EA 642.09 USD
220021-261
Anti-BTK Mouse Monoclonal Antibody [clone: 566928]
Antibodies
Type: Primary
Antigen: BTK
Clonality: Monoclonal
Clone: 566928
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR