Anti-SIGLEC12 Rat Monoclonal Antibody [clone: 750620]

Supplier: R & D Systems
MAB5806
102886-120EA 616.06 USD
102886-120
Anti-SIGLEC12 Rat Monoclonal Antibody [clone: 750620]
Antibodies
Type: Primary
Antigen: SIGLEC12
Clonality: Monoclonal
Clone: 750620
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR