Anti-TIMP1 Mouse Monoclonal Antibody [clone: 150911]

Supplier: R & D Systems
MAB580
220021-258EA 763.57 USD
220021-258
Anti-TIMP1 Mouse Monoclonal Antibody [clone: 150911]
Antibodies
Type: Primary
Antigen: TIMP1
Clonality: Monoclonal
Clone: 150911
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR