Anti-NT5E Mouse Monoclonal Antibody [clone: 606112]

Supplier: R & D Systems
MAB5795
102886-118EA 600.15 USD
102886-118
Anti-NT5E Mouse Monoclonal Antibody [clone: 606112]
Antibodies
Type: Primary
Antigen: NT5E
Clonality: Monoclonal
Clone: 606112
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR