Anti-TSHB Mouse Monoclonal Antibody [clone: 512908]

Supplier: R & D Systems
MAB57941
102886-116EA 594.37 USD
102886-116
Anti-TSHB Mouse Monoclonal Antibody [clone: 512908]
Antibodies
Type: Primary
Antigen: TSHB
Clonality: Monoclonal
Clone: 512908
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR