Anti-CD164 Mouse Monoclonal Antibody [clone: 502021]

Supplier: R & D Systems
MAB5790
220021-257EA 577.02 USD
220021-257
Anti-CD164 Mouse Monoclonal Antibody [clone: 502021]
Antibodies
Type: Primary
Antigen: CD164
Clonality: Monoclonal
Clone: 502021
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR