Anti-ELF3 Mouse Monoclonal Antibody [clone: 662516]

Supplier: R & D Systems
MAB57871
102886-114EA 561.11 USD
102886-114
Anti-ELF3 Mouse Monoclonal Antibody [clone: 662516]
Antibodies
Type: Primary
Antigen: ELF3
Clonality: Monoclonal
Clone: 662516
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR