Anti-ELF3 Mouse Monoclonal Antibody [clone: 662533]

Supplier: R & D Systems
MAB5787
102886-112EA 577.02 USD
102886-112
Anti-ELF3 Mouse Monoclonal Antibody [clone: 662533]
Antibodies
Type: Primary
Antigen: ELF3
Clonality: Monoclonal
Clone: 662533
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR