Anti-IGFBP5 Rat Monoclonal Antibody [clone: 127535]

Supplier: R & D Systems
MAB578
220021-255EA 772.25 USD
220021-255
Anti-IGFBP5 Rat Monoclonal Antibody [clone: 127535]
Antibodies
Type: Primary
Antigen: IGFBP5
Clonality: Monoclonal
Clone: 127535
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR