Anti-ACVR1C Mouse Monoclonal Antibody [clone: 11831]

Supplier: R & D Systems
MAB577
220021-253EA 763.57 USD
220021-253
Anti-ACVR1C Mouse Monoclonal Antibody [clone: 11831]
Antibodies
Type: Primary
Antigen: ACVR1C
Clonality: Monoclonal
Clone: 11831
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR