Anti-HTR2C Mouse Monoclonal Antibody [clone: 496214]

Supplier: R & D Systems
MAB5764
102886-110EA 577.02 USD
102886-110
Anti-HTR2C Mouse Monoclonal Antibody [clone: 496214]
Antibodies
Type: Primary
Antigen: HTR2C
Clonality: Monoclonal
Clone: 496214
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR