Anti-SELL Rat Monoclonal Antibody [clone: 95218]

Supplier: R & D Systems
MAB5761
220021-249EA 772.25 USD
220021-249
Anti-SELL Rat Monoclonal Antibody [clone: 95218]
Antibodies
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: 95218
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR