Anti-SELL Rat Monoclonal Antibody [clone: 95205]

Supplier: R & D Systems
MAB576
220021-248EA 763.57 USD
220021-248
Anti-SELL Rat Monoclonal Antibody [clone: 95205]
Antibodies
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: 95205
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR