Anti-ING1 Mouse Monoclonal Antibody [clone: 585922]

Supplier: R & D Systems
MAB57581
102886-108EA 577.02 USD
102886-108
Anti-ING1 Mouse Monoclonal Antibody [clone: 585922]
Antibodies
Type: Primary
Antigen: ING1
Clonality: Monoclonal
Clone: 585922
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR