Anti-ING1 Mouse Monoclonal Antibody [clone: 585915]

Supplier: R & D Systems
MAB5758
102886-106EA 561.11 USD
102886-106
Anti-ING1 Mouse Monoclonal Antibody [clone: 585915]
Antibodies
Type: Primary
Antigen: ING1
Clonality: Monoclonal
Clone: 585915
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR