Anti-CSH1 Mouse Monoclonal Antibody [clone: 658230]

Supplier: R & D Systems
MAB5757
102886-104EA 594.37 USD
102886-104
Anti-CSH1 Mouse Monoclonal Antibody [clone: 658230]
Antibodies
Type: Primary
Antigen: CSH1
Clonality: Monoclonal
Clone: 658230
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR