Anti-ENPP3 Mouse Monoclonal Antibody [clone: NP4D6]

Supplier: R & D Systems
MAB5756
220021-247EA 594.37 USD
220021-247
Anti-ENPP3 Mouse Monoclonal Antibody [clone: NP4D6]
Antibodies
Type: Primary
Antigen: ENPP3
Clonality: Monoclonal
Clone: NP4D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR