Anti-GDF3 Mouse Monoclonal Antibody [clone: 741614]

Supplier: R & D Systems
MAB5754
102886-102EA 566.89 USD
102886-102
Anti-GDF3 Mouse Monoclonal Antibody [clone: 741614]
Antibodies
Type: Primary
Antigen Symbol: GDF3
Antigen Name: growth differentiation factor 3
Antigen Synonyms: KFS3, MCOP7, MCOPCB6
Clonality: Monoclonal
Clone: 741614
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR