Anti-SELE Rat Monoclonal Antibody [clone: 96403]

Supplier: R & D Systems
MAB575
220021-245EA 763.57 USD
220021-245
Anti-SELE Rat Monoclonal Antibody [clone: 96403]
Antibodies
Type: Primary
Antigen: SELE
Clonality: Monoclonal
Clone: 96403
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR