Anti-TF Mouse Monoclonal Antibody [clone: 507506]

Supplier: R & D Systems
MAB5746
220021-244EA 600.15 USD
220021-244
Anti-TF Mouse Monoclonal Antibody [clone: 507506]
Antibodies
Type: Primary
Antigen: TF
Clonality: Monoclonal
Clone: 507506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR