Anti-SNCG Mouse Monoclonal Antibody [clone: 514304]

Supplier: R & D Systems
MAB5745
220021-243EA 584.25 USD
220021-243
Anti-SNCG Mouse Monoclonal Antibody [clone: 514304]
Antibodies
Type: Primary
Antigen: SNCG
Clonality: Monoclonal
Clone: 514304
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR