Anti-LGALS14 Mouse Monoclonal Antibody [clone: 511124]

Supplier: R & D Systems
MAB5744
220021-242EA 577.02 USD
220021-242
Anti-LGALS14 Mouse Monoclonal Antibody [clone: 511124]
Antibodies
Type: Primary
Antigen: LGALS14
Clonality: Monoclonal
Clone: 511124
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR