Anti-CD27 Rat Monoclonal Antibody [clone: 137915]

Supplier: R & D Systems
MAB5741
220021-241EA 763.57 USD
220021-241
Anti-CD27 Rat Monoclonal Antibody [clone: 137915]
Antibodies
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: 137915
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR