Anti-CD27 Rat Monoclonal Antibody [clone: 137910]

Supplier: R & D Systems
MAB574
220021-240EA 577.02 USD
220021-240
Anti-CD27 Rat Monoclonal Antibody [clone: 137910]
Antibodies
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: 137910
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR