Anti-CAV1 Mouse Monoclonal Antibody [clone: 7C8]

Supplier: R & D Systems
MAB5736
220021-239EA 633.42 USD
220021-239
Anti-CAV1 Mouse Monoclonal Antibody [clone: 7C8]
Antibodies
Type: Primary
Antigen: CAV1
Clonality: Monoclonal
Clone: 7C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR