Anti-IL1RL2 Mouse Monoclonal Antibody [clone: 131011]

Supplier: R & D Systems
MAB573
220021-238EA 763.57 USD
220021-238
Anti-IL1RL2 Mouse Monoclonal Antibody [clone: 131011]
Antibodies
Type: Primary
Antigen: IL1RL2
Clonality: Monoclonal
Clone: 131011
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR