Anti-THAP11 Mouse Monoclonal Antibody [clone: 605383]

Supplier: R & D Systems
MAB5727
102886-096EA 594.37 USD
102886-096
Anti-THAP11 Mouse Monoclonal Antibody [clone: 605383]
Antibodies
Simple Western

Type: Primary
Antigen: THAP11
Clonality: Monoclonal
Clone: 605383
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR