Anti-CXCL11 Rat Monoclonal Antibody [clone: 131327]

Supplier: R & D Systems
MAB572
220021-237EA 763.57 USD
220021-237
Anti-CXCL11 Rat Monoclonal Antibody [clone: 131327]
Antibodies
Type: Primary
Antigen: CXCL11
Clonality: Monoclonal
Clone: 131327
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR