Anti-ESR1 Mouse Monoclonal Antibody [clone: 681118]

Supplier: R & D Systems
MAB57151
102886-090EA 594.37 USD
102886-090
Anti-ESR1 Mouse Monoclonal Antibody [clone: 681118]
Antibodies
Simple Western

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: 681118
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR