Anti-ESR1 Mouse Monoclonal Antibody [clone: 585112]

Supplier: R & D Systems
MAB5715
102886-088EA 577.02 USD
102886-088
Anti-ESR1 Mouse Monoclonal Antibody [clone: 585112]
Antibodies
Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: 585112
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR