Anti-EPM2A Mouse Monoclonal Antibody [clone: 523435]

Supplier: R & D Systems
MAB5714
220021-236EA 561.11 USD
220021-236
Anti-EPM2A Mouse Monoclonal Antibody [clone: 523435]
Antibodies
Type: Primary
Antigen: EPM2A
Clonality: Monoclonal
Clone: 523435
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR