Anti-RAP2B Mouse Monoclonal Antibody [clone: 455517]

Supplier: R & D Systems
MAB5710
220021-235EA 577.02 USD
220021-235
Anti-RAP2B Mouse Monoclonal Antibody [clone: 455517]
Antibodies
Type: Primary
Antigen: RAP2B
Clonality: Monoclonal
Clone: 455517
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR