Anti-PTH1R Mouse Monoclonal Antibody [clone: 734630]

Supplier: R & D Systems
MAB5709
102886-086EA 594.37 USD
102886-086
Anti-PTH1R Mouse Monoclonal Antibody [clone: 734630]
Antibodies
Type: Primary
Antigen: PTH1R
Clonality: Monoclonal
Clone: 734630
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR