Anti-COL18A1 Rat Monoclonal Antibody [clone: 91318]

Supplier: R & D Systems
MAB570
220021-232EA 763.57 USD
220021-232
Anti-COL18A1 Rat Monoclonal Antibody [clone: 91318]
Antibodies
Type: Primary
Antigen: COL18A1
Clonality: Monoclonal
Clone: 91318
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR