Anti-SORL1 Mouse Monoclonal Antibody [clone: 525122]

Supplier: R & D Systems
MAB5699
220021-231EA 577.02 USD
220021-231
Anti-SORL1 Mouse Monoclonal Antibody [clone: 525122]
Antibodies
Type: Primary
Antigen: SORL1
Clonality: Monoclonal
Clone: 525122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR