Anti-FOLR2 Mouse Monoclonal Antibody [clone: 583121]

Supplier: R & D Systems
MAB5697
102886-084EA 594.37 USD
102886-084
Anti-FOLR2 Mouse Monoclonal Antibody [clone: 583121]
Antibodies
Type: Primary
Antigen Symbol: FOLR2
Antigen Name: folate receptor 2 (fetal)
Antigen Synonyms: BETA-HFR, FBP, FBP/PL-1, FR-BETA, FR-P3
Clonality: Monoclonal
Clone: 583121
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR