Anti-SOCS3 Mouse Monoclonal Antibody [clone: 516919]

Supplier: R & D Systems
MAB5696
220021-230EA 600.15 USD
220021-230
Anti-SOCS3 Mouse Monoclonal Antibody [clone: 516919]
Antibodies
Type: Primary
Antigen: SOCS3
Clonality: Monoclonal
Clone: 516919
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR