Anti-MET Mouse Monoclonal Antibody [clone: 527125]

Supplier: R & D Systems
MAB5694
220021-228EA 577.02 USD
220021-228
Anti-MET Mouse Monoclonal Antibody [clone: 527125]
Antibodies
Type: Primary
Antigen: MET
Clonality: Monoclonal
Clone: 527125
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR