Anti-CD96 Rat Monoclonal Antibody [clone: 630612]

Supplier: R & D Systems
MAB5690
102886-082EA 577.02 USD
102886-082
Anti-CD96 Rat Monoclonal Antibody [clone: 630612]
Antibodies
Type: Primary
Antigen: CD96
Clonality: Monoclonal
Clone: 630612
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR