Anti-EFNA4 Rat Monoclonal Antibody [clone: 132416]

Supplier: R & D Systems
MAB569
220021-227EA 763.57 USD
220021-227
Anti-EFNA4 Rat Monoclonal Antibody [clone: 132416]
Antibodies
Type: Primary
Antigen: EFNA4
Clonality: Monoclonal
Clone: 132416
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR