Anti-PKN2 Mouse Monoclonal Antibody [clone: 509105]

Supplier: R & D Systems
MAB5686
220021-226EA 594.37 USD
220021-226
Anti-PKN2 Mouse Monoclonal Antibody [clone: 509105]
Antibodies
Type: Primary
Antigen: PKN2
Clonality: Monoclonal
Clone: 509105
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR