Anti-ITGA1 Mouse Monoclonal Antibody [clone: 639508]

Supplier: R & D Systems
MAB5676
102886-080EA 594.37 USD
102886-080
Anti-ITGA1 Mouse Monoclonal Antibody [clone: 639508]
Antibodies
Type: Primary
Antigen: ITGA1
Clonality: Monoclonal
Clone: 639508
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR