Anti-L1CAM Rat Monoclonal Antibody [clone: 555]

Supplier: R & D Systems
MAB5674
220021-224EA 594.37 USD
220021-224
Anti-L1CAM Rat Monoclonal Antibody [clone: 555]
Antibodies
Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone: 555
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR