Anti-TNFA Mouse Monoclonal Antibody [clone: 610325]

Supplier: R & D Systems
MAB56701
102886-078EA 594.37 USD
102886-078
Anti-TNFA Mouse Monoclonal Antibody [clone: 610325]
Antibodies
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: 610325
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR