Anti-TNFA Mouse Monoclonal Antibody [clone: 610331]

Supplier: R & D Systems
MAB5670
102886-076EA 763.57 USD
102886-076
Anti-TNFA Mouse Monoclonal Antibody [clone: 610331]
Antibodies
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: 610331
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR